Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: RSPH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286506.1 | NP_001273435.1 | 8 | 1263 | ||
NM_080860.3 | NP_543136.1 | 9 | 1377 | |||
XM_005261208.2 | XP_005261265.1 | 7 | 1017 | |||
XM_011529786.1 | XP_011528088.1 | 8 | 1197 | |||
GenBank mRNA | AB006536.1 | - | 9 | 1377 |