Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCFD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171506.2 | NP_001164977.1 | 3, 4 | 395 | ||
NM_001171507.2 | NP_001164978.1 | 2, 3 | 287 | |||
NM_001171508.2 | NP_001164979.1 | 2, 3 | 439 | |||
NM_139279.5 | NP_644808.1 | 2, 3 | 301 | |||
GenBank mRNA | AF475284.1 | - | 2, 3 | 207 | ||
AF537214.1 | - | 2, 3 | 238 | |||
AK292127.1 | - | 2, 3 | 316 | |||
AL833900.1 | - | 2, 3 | 236 | |||
BC040357.2 | - | 2, 3 | 227 | 3925166 | ||
CF994192.1 | - | 3, 4 | 339 | |||
DA226366.1 | - | 3, 4 | 317 | |||
DA324623.1 | - | 2, 3 | 292 | |||
DA404031.1 | - | 2, 3 | 287 |