Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACBD5 SIRNA Assays ›
Gene Symbol:
ACBD5
Gene Name:
acyl-CoA binding domain containing 5
Gene Aliases:
-
Chromosome Location:
Chr.10: 27182624 - 27248674 on Build GRCh38
UniGene ID:
Hs.530597
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
131805
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ACBD5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001042473.3
NP_001035938.1
9, 10
1278
NM_001271512.3
NP_001258441.1
8, 9
1592
NM_001301251.1
NP_001288180.1
8, 9
1157
NM_001301252.1
NP_001288181.1
7, 8
1471
NM_001301253.1
NP_001288182.1
8, 9
1147
NM_001301254.1
NP_001288183.1
6, 7
1267
NM_145698.4
NP_663736.2
9, 10
1403
XM_006717528.3
XP_006717591.3
10, 11
7836
XM_006717530.3
XP_006717593.1
10, 11
1435
XM_006717531.2
XP_006717594.1
10, 11
1445
XM_006717535.2
XP_006717598.1
10, 11
1302
XM_017016884.1
XP_016872373.1
10, 11
7836
XM_017016885.1
XP_016872374.1
9, 10
7803
XM_017016886.1
XP_016872375.1
9, 10
7803
XM_017016887.1
XP_016872376.1
10, 11
1366
XM_017016888.1
XP_016872377.1
10, 11
1431
XM_017016889.1
XP_016872378.1
9, 10
1333
XM_017016890.1
XP_016872379.1
9, 10
7632
XM_017016891.1
XP_016872380.1
9, 10
1398
XM_017016892.1
XP_016872381.1
9, 10
1319
XM_017016893.1
XP_016872382.1
8, 9
7599
XM_017016894.1
XP_016872383.1
8, 9
7599
XM_017016895.1
XP_016872384.1
10, 11
1302
XM_017016896.1
XP_016872385.1
9, 10
1270
XM_017016897.1
XP_016872386.1
9, 10
1227
XM_017016898.1
XP_016872387.1
8, 9
1194
XM_017016899.1
XP_016872388.1
8, 9
1194
XM_017016900.1
XP_016872389.1
9, 10
1220
XM_017016901.1
XP_016872390.1
9, 10
1200
XM_017016902.1
XP_016872391.1
9, 10
1219
XM_017016903.1
XP_016872392.1
9, 10
1204
XM_017016904.1
XP_016872393.1
8, 9
1199
XM_017016905.1
XP_016872394.1
8, 9
1185
XM_017016906.1
XP_016872395.1
8, 9
1066
XM_017016907.1
XP_016872396.1
8, 9
1199
XM_017016908.1
XP_016872397.1
8, 9
1188
GenBank mRNA
AB082527.1
-
8, 9
1106
AF505653.1
-
8, 9
1144
AK057469.1
-
9, 10
1270
AK294510.1
-
6, 7
1185
AK295031.1
-
8, 9
1229
AK304842.1
-
7, 8
1122
AL133064.1
-
3, 4
486
BC030555.1
-
9, 10
1307
5218685
Back To Top
More Information
Panther Classification:
Molecular Function -
transfer/carrier protein
Gene Ontology Categories:
Function(s)
Process(es)
transport
macroautophagy
pexophagy
aggrephagy
fatty-acyl-CoA binding
molecular_function
lipid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System