Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127372.2 | NP_001120844.1 | 5 | 838 | ||
NM_001289971.1 | NP_001276900.1 | 5 | 829 | |||
NM_001289972.1 | NP_001276901.1 | 5 | 957 | |||
NM_003428.5 | NP_003419.3 | 5 | 822 | |||
XM_005266185.1 | XP_005266242.1 | 5 | 992 | |||
XM_005266186.1 | XP_005266243.1 | 6 | 728 | |||
XM_011534832.1 | XP_011533134.1 | 5 | 772 | |||
GenBank mRNA | AK314232.1 | - | 5 | 600 | ||
BC036656.1 | - | 5 | 944 | 4828620 | ||
BC048350.1 | - | 4 | 406 | 5555056 | ||
BC062552.1 | - | 5 | 598 | 6536224 | ||
M27878.1 | - | 5 | 730 | |||
X60156.2 | - | 4 | 568 |