Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDK5RAP2 SIRNA Assays ›
          Gene Symbol:
          CDK5RAP2
          Gene Name:
          CDK5 regulatory subunit associated protein 2
          Gene Aliases:
          C48, Cep215, MCPH3
          Chromosome Location:
          Chr.9: 120388869 - 120580170 on Build GRCh38
          UniGene ID:
          Hs.269560
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 132392

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDK5RAP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001011649.2 NP_001011649.1 10 1155
          NM_001272039.1 NP_001258968.1 10 1155
          NM_018249.5 NP_060719.4 10 1155
          NR_073554.1 10 1155
          NR_073555.1 10 1155
          NR_073556.1 10 1152
          NR_073557.1 10 1155
          NR_073558.1 10 1152
          XM_006717182.1 XP_006717245.1 10 1144
          XM_006717185.1 XP_006717248.1 10 1144
          XM_011518860.1 XP_011517162.1 10 1144
          XM_017014921.1 XP_016870410.1 10 1144
          XM_017014922.1 XP_016870411.1 4 373
          XM_017014923.1 XP_016870412.1 10 1144
          XM_017014924.1 XP_016870413.1 1 49
          GenBank mRNA AB046853.2 - 10 1124
          AF448860.1 - 10 1105
          AK122913.1 - 10 1144
          AL833481.1 - 10 1143
          BC136275.1 - 10 990 9020262
          BC143762.1 - 10 990 9052276
          BC146782.1 - 10 1124 9007237
          BX537421.1 - 10 1140
          BX537708.1 - 10 1144
          BX537759.1 - 10 1143
          BX640896.1 - 4 367
          CR936622.1 - 3 285
          CR936717.1 - 3 285
          KP294527.1 - 10 1155

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          establishment of mitotic spindle orientation
          microtubule cytoskeleton organization
          microtubule bundle formation
          chromosome segregation
          centriole replication
          brain development
          neurogenesis
          microtubule organizing center organization
          regulation of neuron differentiation
          negative regulation of neuron differentiation
          positive regulation of transcription, DNA-templated
          negative regulation of centriole replication
          centrosome organization
          regulation of spindle checkpoint
          protein binding
          calmodulin binding
          microtubule binding
          tubulin binding
          protein kinase binding
          protein complex binding
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nq57x:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0