Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TDP1 SIRNA Assays ›
          Gene Symbol:
          TDP1
          Gene Name:
          tyrosyl-DNA phosphodiesterase 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 89954935 - 90044764 on Build GRCh38
          UniGene ID:
          Hs.209945
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 132432

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TDP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001008744.1 NP_001008744.1 8 1092
          NM_018319.3 NP_060789.2 9 1315
          XM_005267847.2 XP_005267904.1 9 1989
          XM_005267848.2 XP_005267905.1 8 1333
          XM_006720197.3 XP_006720260.1 9 1556
          XM_006720198.3 XP_006720261.1 8 1211
          XM_006720199.1 XP_006720262.1 8 697
          XM_011536942.2 XP_011535244.1 8 1732
          XM_011536943.2 XP_011535245.1 8 1125
          XM_011536944.1 XP_011535246.1 9 1316
          XM_017021439.1 XP_016876928.1 9 1223
          XM_017021440.1 XP_016876929.1 8 1115
          XM_017021441.1 XP_016876930.1 8 1304
          XM_017021442.1 XP_016876931.1 6 941
          XM_017021443.1 XP_016876932.1 6 440
          XM_017021444.1 XP_016876933.1 6 441
          XM_017021445.1 XP_016876934.1 6 941
          XM_017021446.1 XP_016876935.1 7 747
          GenBank mRNA AF182002.1 - 7 1001
          AK001952.1 - 8 1056
          AK093235.1 - 10 2123
          AK315549.1 - 8 1040
          BC006083.1 - 10 1422
          BC015474.1 - 9 1266 3900062
          BC107877.1 - 9 1319 6597752
          BX161451.1 - 8 1227

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phosphodiesterase

          Gene Ontology Categories:

          Function(s) Process(es)

          single strand break repair
          DNA repair
          double-strand break repair
          nucleic acid phosphodiester bond hydrolysis
          double-stranded DNA binding
          single-stranded DNA binding
          exonuclease activity
          protein binding
          3'-tyrosyl-DNA phosphodiesterase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vrbfv:80/100.66.76.55:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0