Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LAPTM4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018407.4 | NP_060877.3 | 7 | 1918 | ||
GenBank mRNA | AK075326.1 | - | 7 | 1680 | ||
AL136942.1 | - | 7 | 1672 | |||
AY057051.2 | - | 7 | 1918 | |||
AY261384.1 | - | 7 | 1937 | |||
BC014129.2 | - | 7 | 1674 | 4581284 | ||
BC031021.1 | - | 7 | 1699 | 4690368 | ||
BC038117.1 | - | 7 | 2466 | 5264567 |