Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018451.4 | NP_060921.3 | 8 | 2975 | ||
NR_047594.1 | 8 | 2975 | ||||
NR_047595.1 | 8 | 2975 | ||||
XM_011535149.2 | XP_011533451.1 | 8 | 2975 | |||
XM_011535150.2 | XP_011533452.1 | 8 | 2975 | |||
XM_017020673.1 | XP_016876162.1 | 8 | 2975 | |||
GenBank mRNA | AF139625.1 | - | 8 | 2990 | ||
AF141343.1 | - | 1 | 24 | |||
AL833182.1 | - | 8 | 2922 | |||
BC113110.1 | - | 8 | 2911 | 7961385 | ||
BC113111.1 | - | 8 | 2912 | 7961390 | ||
BC113662.1 | - | 8 | 2862 | 8322714 | ||
BC113664.1 | - | 8 | 2862 | 8322716 | ||
BC143757.1 | - | 8 | 2862 | |||
BC143758.1 | - | 8 | 2862 |