Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ACSS2 SIRNA Assays ›
          Gene Symbol:
          ACSS2
          Gene Name:
          acyl-CoA synthetase short-chain family member 2
          Gene Aliases:
          ACAS2, ACECS, ACS, ACSA, dJ1161H23.1
          Chromosome Location:
          Chr.20: 34874942 - 34927966 on Build GRCh38
          UniGene ID:
          Hs.517034
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 132614

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ACSS2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001076552.2 NP_001070020.2 9, 10 1135
          NM_001242393.1 NP_001229322.1 8, 9 972
          NM_018677.3 NP_061147.1 8, 9 1096
          XM_005260455.1 XP_005260512.1 9, 10 1135
          XM_005260456.1 XP_005260513.1 8, 9 1096
          XM_006723826.1 XP_006723889.1 9, 10 1011
          XM_011528905.1 XP_011527207.1 10, 11 1127
          XM_011528906.1 XP_011527208.1 10, 11 1127
          XM_011528907.1 XP_011527209.1 9, 10 1088
          XM_011528908.1 XP_011527210.1 9, 10 1088
          XM_011528909.2 XP_011527211.1 10, 11 1013
          XM_011528911.1 XP_011527213.1 9, 10 1022
          XM_011528912.2 XP_011527214.1 10, 11 1019
          GenBank mRNA AF263614.1 - 8, 9 975
          AK000162.1 - 8, 9 911
          AK000188.1 - 8, 9 1049
          AK022608.1 - 2, 3 208
          AK025990.1 - 8, 9 1031
          AK092281.1 - 9, 10 1047
          AK098026.1 - 3, 4 571
          AL359946.1 - 7, 8 805
          BC010141.2 - 5, 6 475
          BC012172.1 - 8, 9 1019 4643236
          BC073846.1 - 8, 9 860
          BC098422.1 - 7, 8 691 5798672
          CR749716.1 - 7, 8 805

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          ethanol oxidation
          lipid biosynthetic process
          acetate biosynthetic process
          acetyl-CoA biosynthetic process from acetate
          propionate biosynthetic process
          acetate-CoA ligase activity
          ATP binding
          AMP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline