Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCHCR1 SIRNA Assays ›
          Gene Symbol:
          CCHCR1
          Gene Name:
          coiled-coil alpha-helical rod protein 1
          Gene Aliases:
          C6orf18, HCR, SBP
          Chromosome Location:
          Chr.6: 31142439 - 31158238 on Build GRCh38
          UniGene ID:
          Hs.485075
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 132824

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCHCR1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001105563.1 NP_001099033.1 3 543
          NM_001105564.1 NP_001099034.1 3 651
          NM_019052.3 NP_061925.2 3 318
          XM_011514702.2 XP_011513004.1 3 577
          XM_011514703.1 XP_011513005.1 4 390
          XM_011514704.2 XP_011513006.1 3 179
          XM_011514705.1 XP_011513007.1 3 288
          XM_017010961.1 XP_016866450.1 3 685
          XM_017010962.1 XP_016866451.1 4 437
          XM_017010963.1 XP_016866452.1 4 551
          XM_017010964.1 XP_016866453.1 3 315
          XM_017010965.1 XP_016866454.1 3 377
          XM_017010966.1 XP_016866455.1 3 398
          XM_017010967.1 XP_016866456.1 3 501
          XM_017010968.1 XP_016866457.1 3 270
          XM_017010969.1 XP_016866458.1 3 177
          XM_017010970.1 XP_016866459.1 3 267
          GenBank mRNA AB029331.1 - 1 139
          AB112474.1 - 3 274
          AB112475.1 - 3 625
          AF216493.1 - 1 139
          AK000204.1 - 3 274
          AK000217.1 - 3 275
          AK289446.1 - 3 266
          AK295494.1 - 3 338
          AK310626.1 - 3 336
          AY029160.1 - 2 195
          BC110534.1 - 3 361 40035162
          BC110535.1 - 3 474 40035164
          DC357362.1 - 3 318

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s)

          protein export from nucleus
          multicellular organism development
          cell differentiation

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-85894:80/100.66.79.23:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0