Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PARD3 SIRNA Assays ›
          Gene Symbol:
          PARD3
          Gene Name:
          par-3 family cell polarity regulator
          Gene Aliases:
          ASIP, Baz, PAR3, PAR3alpha, PARD-3, PARD3A, PPP1R118, SE2-5L16, SE2-5LT1, SE2-5T2
          Chromosome Location:
          Chr.10: 34109560 - 34815325 on Build GRCh38
          UniGene ID:
          Hs.131489
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 132895

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PARD3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001184785.1 NP_001171714.1 10, 11 1877
          NM_001184786.1 NP_001171715.1 10, 11 1877
          NM_001184787.1 NP_001171716.1 10, 11 1877
          NM_001184788.1 NP_001171717.1 10, 11 1877
          NM_001184789.1 NP_001171718.1 10, 11 1877
          NM_001184790.1 NP_001171719.1 9, 10 1745
          NM_001184791.1 NP_001171720.1 9, 10 1745
          NM_001184792.1 NP_001171721.1 10, 11 1877
          NM_001184793.1 NP_001171722.1 10, 11 1877
          NM_001184794.1 NP_001171723.1 10, 11 1877
          NM_019619.3 NP_062565.2 10, 11 1877
          XM_005252528.4 XP_005252585.1 10, 11 1868
          XM_005252530.4 XP_005252587.1 10, 11 1868
          XM_005252531.4 XP_005252588.1 10, 11 1868
          XM_005252532.4 XP_005252589.1 9, 10 1522
          XM_005252534.4 XP_005252591.1 10, 11 1868
          XM_005252535.4 XP_005252592.1 10, 11 1868
          XM_005252536.4 XP_005252593.1 10, 11 1868
          XM_011519569.2 XP_011517871.1 10, 11 1868
          XM_011519570.2 XP_011517872.1 10, 11 1868
          XM_011519571.2 XP_011517873.1 10, 11 1868
          XM_011519572.2 XP_011517874.1 10, 11 1868
          XM_011519573.2 XP_011517875.1 10, 11 1868
          XM_011519575.2 XP_011517877.1 9, 10 1552
          XM_011519576.2 XP_011517878.1 9, 10 1523
          XM_011519578.2 XP_011517880.1 9, 10 1740
          XM_011519579.2 XP_011517881.1 8, 9 1421
          XM_011519581.2 XP_011517883.1 10, 11 1868
          XM_011519583.2 XP_011517885.1 10, 11 1868
          XM_011519584.2 XP_011517886.1 10, 11 1868
          XM_017016430.1 XP_016871919.1 10, 11 1868
          GenBank mRNA AB073671.1 - 9, 10 1415
          AF177228.1 - 3, 4 613
          AF196185.1 - 10, 11 1834
          AF196186.1 - 10, 11 1834
          AF252293.1 - 9, 10 1415
          AF332592.1 - 10, 11 1547
          AF332593.1 - 10, 11 1547
          AF454057.1 - 8, 9 1254
          AF454058.1 - 9, 10 1759
          AF454059.1 - 6, 7 880
          AF467002.1 - 10, 11 1547
          AF467003.1 - 10, 11 1547
          AF467004.1 - 10, 11 1547
          AF467005.1 - 9, 10 1415
          AF467006.1 - 10, 11 1547
          AK025892.1 - 2, 3 363
          AK027735.1 - 8, 9 1174
          BC071566.1 - 10, 11 1824 30344732

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein complex assembly
          protein targeting to membrane
          cell cycle
          establishment or maintenance of cell polarity
          transforming growth factor beta receptor signaling pathway
          protein kinase C-activating G-protein coupled receptor signaling pathway
          axonogenesis
          asymmetric cell division
          negative regulation of peptidyl-threonine phosphorylation
          myelination in peripheral nervous system
          positive regulation of myelination
          regulation of cellular localization
          bicellular tight junction assembly
          establishment of epithelial cell polarity
          protein binding
          phosphatidylinositol-4,5-bisphosphate binding
          phosphatidylinositol-3,4,5-trisphosphate binding
          protein phosphatase binding
          phosphatidylinositol-3-phosphate binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0