Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PMEPA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001255976.1 | NP_001242905.1 | 4 | 1821 | ||
NM_020182.4 | NP_064567.2 | 4 | 2092 | |||
NM_199169.2 | NP_954638.1 | 4 | 1745 | |||
NM_199170.2 | NP_954639.1 | 4 | 1738 | |||
NM_199171.2 | NP_954640.1 | 4 | 1803 | |||
GenBank mRNA | AF305616.1 | - | 4 | 2019 | ||
AY128643.1 | - | 4 | 1693 |