Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: APMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020531.2 | NP_065392.1 | 7, 8 | 948 | ||
GenBank mRNA | AB033767.1 | - | 7, 8 | 887 | ||
AK025834.1 | - | 3, 4 | 412 | |||
AK026866.1 | - | 4, 5 | 446 | |||
AK291129.1 | - | 7, 8 | 909 | |||
AY359076.1 | - | 6, 7 | 735 | |||
BC003501.1 | - | 7, 8 | 872 | 3530962 |