Search
Search

MKL1| Gene Symbol: MKL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282660.1 | NP_001269589.1 | 5, 6 | 694 | ||
| NM_001282662.1 | NP_001269591.1 | 8, 9 | 1106 | |||
| NM_001318139.1 | NP_001305068.1 | 6, 7 | 831 | |||
| NM_020831.4 | NP_065882.1 | 8, 9 | 1106 | |||
| XM_005261692.1 | XP_005261749.1 | 7, 8 | 911 | |||
| XM_005261694.1 | XP_005261751.1 | 7, 8 | 1001 | |||
| XM_011530283.1 | XP_011528585.1 | 9, 10 | 1214 | |||
| XM_011530285.1 | XP_011528587.1 | 9, 10 | 1125 | |||
| XM_017028888.1 | XP_016884377.1 | 8, 9 | 1189 | |||
| XM_017028889.1 | XP_016884378.1 | 4, 5 | 2189 | |||
| GenBank mRNA | AB037859.2 | - | 8, 9 | 1078 | ||
| AF368061.1 | - | 8, 9 | 1031 | |||
| AJ297257.1 | - | 5, 6 | 688 | |||
| AJ297258.1 | - | 8, 9 | 1088 | |||
| BC115039.1 | - | 8, 9 | 1072 | 5551767 | ||
| BC139730.1 | - | 6, 7 | 745 | |||
| CR456522.1 | - | 5, 6 | 505 | |||
| HG764171.1 | - | 6, 7 | 831 | |||