Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTBP2 SIRNA Assays ›
          Gene Symbol:
          PTBP2
          Gene Name:
          polypyrimidine tract binding protein 2
          Gene Aliases:
          PTBLP, brPTB, nPTB
          Chromosome Location:
          Chr.1: 96721605 - 96815049 on Build GRCh38
          UniGene ID:
          Hs.596061
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 133331

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PTBP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001300985.1 NP_001287914.1 12 1650
          NM_001300986.1 NP_001287915.1 12 1659
          NM_001300987.1 NP_001287916.1 13 1806
          NM_001300988.1 NP_001287917.1 12 1650
          NM_001300989.1 NP_001287918.1 12 1635
          NM_001300990.1 NP_001287919.1 11 1559
          NM_021190.3 NP_067013.1 12 1635
          NR_125356.1 11 1493
          NR_125357.1 11 1601
          XM_011541874.2 XP_011540176.1 12 1483
          XM_011541875.2 XP_011540177.1 12 1481
          XM_011541876.2 XP_011540178.1 12 1513
          XM_017001951.1 XP_016857440.1 11 1513
          GenBank mRNA AB051232.1 - 12 1457
          AB051233.1 - 11 1367
          AF176085.1 - 12 1427
          AF530580.1 - 12 1445
          AF530581.1 - 12 1445
          AF530582.1 - 4 498
          AF530583.1 - 4 513
          AF530584.1 - 4 498
          AF530585.1 - 4 513
          AK022716.1 - 7 941
          AK025333.1 - 2 298
          AK092216.1 - 11 1499
          AK295384.1 - 11 1414
          AK299755.1 - 11 1244
          AK299897.1 - 13 1587
          AK299922.1 - 12 1473
          BC016582.2 - 12 1404 3872881

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splice site selection
          spinal cord development
          cerebellum development
          negative regulation of RNA splicing
          regulation of neural precursor cell proliferation
          nucleotide binding
          mRNA binding
          pre-mRNA binding
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-xmktn:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0