Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Food and Beverage
Lab Solutions
Pharma and Biopharma
Real-Time PCR
Semiconductor Analysis
Clinical and Diagnostics
Digital Solutions
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
See all services
Help and Support
Order Help
Digital Solutions
Product Support
Technical Information
Training and Education
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYTL2 SIRNA Assays ›
Gene Symbol:
SYTL2
Gene Name:
synaptotagmin like 2
Gene Aliases:
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
Chromosome Location:
Chr.11: 85694221 - 85811182 on Build GRCh38
UniGene ID:
Hs.369520
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133767
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYTL2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001162951.2
NP_001156423.1
14
2527
NM_001162952.2
NP_001156424.1
6
1057
NM_001162953.2
NP_001156425.1
14
2530
NM_001289608.1
NP_001276537.1
15
2336
NM_001289609.1
NP_001276538.1
7
1170
NM_001289610.1
NP_001276539.1
7
741
NM_032943.4
NP_116561.1
15
2706
NM_206929.3
NP_996812.1
8
861
NM_206930.3
NP_996813.1
5
1034
XM_005274057.1
XP_005274114.1
14
6166
XM_005274060.1
XP_005274117.1
15
2350
XM_005274061.1
XP_005274118.1
15
2347
XM_005274062.2
XP_005274119.1
16
2771
XM_005274063.1
XP_005274120.1
14
2230
XM_005274066.1
XP_005274123.1
13
2179
XM_005274067.4
XP_005274124.1
7
2473
XM_011545105.1
XP_011543407.1
15
6214
XM_011545106.1
XP_011543408.1
15
6211
XM_011545107.2
XP_011543409.1
16
6635
XM_011545108.1
XP_011543410.1
14
6094
XM_011545109.2
XP_011543411.1
15
6196
XM_011545114.2
XP_011543416.1
6
2379
XM_011545115.1
XP_011543417.1
4
473
XM_011545116.2
XP_011543418.1
5
2258
XM_017017933.1
XP_016873422.1
14
6163
XM_017017934.1
XP_016873423.1
16
6632
XM_017017935.1
XP_016873424.1
12
6010
XM_017017936.1
XP_016873425.1
12
5708
XM_017017937.1
XP_016873426.1
15
2251
XM_017017938.1
XP_016873427.1
14
2209
XM_017017939.1
XP_016873428.1
8
2517
XM_017017940.1
XP_016873429.1
7
2469
GenBank mRNA
AB046817.1
-
15
2717
AK024872.1
-
5
1198
AK074737.1
-
8
818
AK092121.1
-
5
852
AK124754.1
-
8
2442
AK127504.1
-
9
3405
AK131365.1
-
8
1334
AK222707.1
-
5
1198
AK222710.1
-
5
1198
AK296224.1
-
15
2318
AK296740.1
-
12
2266
AK298604.1
-
7
659
AL834422.1
-
2
96
AY386362.1
-
8
4721
BC015540.2
-
5
528
BC068495.1
-
7
895
BC110315.1
-
6
586
BC136450.1
-
14
2462
9020439
BC144114.1
-
14
2462
9052634
BC144115.1
-
14
2459
9052635
BX647671.1
-
9
3157
BX648068.1
-
8
2603
EU035829.1
-
14
2183
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
intracellular protein transport
exocytosis
vesicle docking involved in exocytosis
vesicle fusion
negative regulation of phosphatase activity
vesicle-mediated transport
regulation of calcium ion-dependent exocytosis
calcium ion-regulated exocytosis of neurotransmitter
positive regulation of mucus secretion
protein localization to plasma membrane
phosphatidylserine binding
calcium ion binding
protein binding
calcium-dependent phospholipid binding
phosphatidylinositol-4,5-bisphosphate binding
Rab GTPase binding
phosphatase binding
syntaxin binding
clathrin binding
neurexin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-74fd55b88b-kd7lp:80/10.239.160.160:80.
git-commit: e4d250c089da5f210ba2ef291b70bc15861d50fd
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.9.0-2024.05.21-1.0