Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CIZ1 SIRNA Assays ›
Gene Symbol:
CIZ1
Gene Name:
CDKN1A interacting zinc finger protein 1
Gene Aliases:
LSFR1, NP94, ZNF356
Chromosome Location:
Chr.9: 128166065 - 128204383 on Build GRCh38
UniGene ID:
Hs.212395
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
133815
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CIZ1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001131015.1
NP_001124487.1
15
2227
NM_001131016.1
NP_001124488.1
14
2355
NM_001131017.1
NP_001124489.1
15
2172
NM_001131018.1
NP_001124490.1
14
2115
NM_001257975.1
NP_001244904.1
15
2525
NM_001257976.1
NP_001244905.1
13
2239
NM_012127.2
NP_036259.2
14
2395
XM_005251888.3
XP_005251945.2
14
2226
XM_005251891.3
XP_005251948.2
15
2058
XM_005251892.3
XP_005251949.2
15
2043
XM_005251893.4
XP_005251950.4
14
1981
XM_006717039.3
XP_006717102.2
13
2134
XM_011518481.1
XP_011516783.1
14
2211
XM_011518482.2
XP_011516784.2
13
2149
XM_011518483.1
XP_011516785.1
13
2266
XM_017014594.1
XP_016870083.1
14
2679
XM_017014595.1
XP_016870084.1
14
2323
XM_017014596.1
XP_016870085.1
15
2170
GenBank mRNA
AB030835.1
-
14
2308
AF159025.1
-
13
2187
AF234161.1
-
12
1958
AK023978.1
-
15
2227
AK292713.1
-
14
2346
AK296363.1
-
15
2526
AK303288.1
-
15
2155
AK303636.1
-
13
2222
AK316393.1
-
12
1951
BC004119.2
-
15
2012
3507940
BC021163.2
-
14
2094
3633796
EF441870.1
-
7
901
Back To Top
More Information
Gene Ontology Categories:
Process(es)
nucleic acid binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0