Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLA2G6 SIRNA Assays ›
Gene Symbol:
PLA2G6
Gene Name:
phospholipase A2 group VI
Gene Aliases:
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
Chromosome Location:
Chr.22: 38111495 - 38192099 on Build GRCh38
UniGene ID:
Hs.170479
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
13444
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLA2G6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001004426.1
NP_001004426.1
4
699
NM_001199562.1
NP_001186491.1
4
654
NM_003560.2
NP_003551.2
4
699
XM_005261764.2
XP_005261821.1
4
674
XM_005261765.1
XP_005261822.1
4
671
XM_005261766.1
XP_005261823.1
4
766
XM_006724332.3
XP_006724395.1
4
675
XM_011530422.1
XP_011528724.1
5
738
XM_011530423.1
XP_011528725.1
5
786
XM_011530424.1
XP_011528726.1
6
910
XM_011530425.1
XP_011528727.1
4
610
XM_011530426.2
XP_011528728.1
4
674
XM_017028981.1
XP_016884470.1
4
766
XM_017028982.1
XP_016884471.1
4
671
XM_017028983.1
XP_016884472.1
6
910
XM_017028984.1
XP_016884473.1
5
786
XM_017028985.1
XP_016884474.1
4
630
XM_017028986.1
XP_016884475.1
4
674
XM_017028987.1
XP_016884476.1
4
674
XM_017028988.1
XP_016884477.1
4
674
GenBank mRNA
AF064594.1
-
4
699
AF102988.1
-
3
563
AF102989.1
-
3
563
AK290825.1
-
4
656
AK291212.1
-
4
685
AK300459.1
-
5
738
AK308308.1
-
3
505
AL080187.1
-
4
699
BC036742.1
-
4
693
5166749
BC051904.1
-
4
635
5175768
CR456543.1
-
4
627
Back To Top
More Information
Panther Classification:
Molecular Function -
phospholipase
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
chemotaxis
positive regulation of cytosolic calcium ion concentration
memory
urinary bladder smooth muscle contraction
lipid catabolic process
antibacterial humoral response
cardiolipin biosynthetic process
response to endoplasmic reticulum stress
positive regulation of insulin secretion involved in cellular response to glucose stimulus
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
Fc-gamma receptor signaling pathway involved in phagocytosis
positive regulation of vasodilation
positive regulation of exocytosis
negative regulation of synaptic transmission, glutamatergic
maternal process involved in female pregnancy
positive regulation of protein kinase C signaling
positive regulation of release of cytochrome c from mitochondria
positive regulation of arachidonic acid secretion
regulation of store-operated calcium channel activity
positive regulation of ceramide biosynthetic process
phospholipase A2 activity
calmodulin binding
hydrolase activity
protein kinase binding
ATP-dependent protein binding
calcium-independent phospholipase A2 activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0