Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: AUTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127231.2 | NP_001120703.1 | 15, 16 | 2897 | ||
NM_015570.3 | NP_056385.1 | 16, 17 | 2969 | |||
XM_005250257.2 | XP_005250314.1 | 9, 10 | 1043 | |||
XM_011516010.2 | XP_011514312.1 | 16, 17 | 3434 | |||
XM_011516011.2 | XP_011514313.1 | 16, 17 | 3431 | |||
XM_011516012.2 | XP_011514314.1 | 15, 16 | 3368 | |||
XM_011516013.2 | XP_011514315.1 | 15, 16 | 3362 | |||
XM_011516014.2 | XP_011514316.1 | 14, 15 | 3332 | |||
XM_011516017.2 | XP_011514319.1 | 15, 16 | 11988 | |||
XM_011516018.2 | XP_011514320.1 | 16, 17 | 1969 | |||
XM_017011951.1 | XP_016867440.1 | 16, 17 | 3434 | |||
GenBank mRNA | AB007902.2 | - | 17, 18 | 2458 | ||
AF326917.1 | - | 16, 17 | 2555 | |||
AK296981.1 | - | 8, 9 | 639 | |||
BC064693.1 | - | 15, 16 | 2905 | 30345831 | ||
JQ670866.1 | - | 15, 16 | 2162 | |||
JQ670867.1 | - | 7, 8 | 566 |