Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PIK3R3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114172.1 | NP_001107644.1 | 5 | 583 | ||
NM_001303427.1 | NP_001290356.1 | 4 | 488 | |||
NM_001303428.1 | NP_001290357.1 | 5 | 427 | |||
NM_001303429.1 | NP_001290358.1 | 4 | 1082 | |||
NM_003629.3 | NP_003620.3 | 4 | 1082 | |||
XM_017002614.1 | XP_016858103.1 | 5 | 460 | |||
GenBank mRNA | AF028785.1 | - | 4 | 1017 | ||
AK302049.1 | - | 4 | 481 | |||
AK313726.1 | - | 5 | 595 | |||
BC005381.1 | - | 4 | 553 | |||
BC021622.1 | - | 4 | 1016 | 3910221 | ||
BX537388.1 | - | 5 | 427 | |||
CR749649.1 | - | 4 | 1003 | |||
D88532.1 | - | 4 | 746 | |||
DB047431.1 | - | 4 | 488 |