Search Thermo Fisher Scientific
Gene Symbol: ELP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288725.1 | NP_001275654.1 | 8 | 1023 | ||
NM_001288726.1 | NP_001275655.1 | 8 | 1020 | |||
NM_019040.4 | NP_061913.3 | 8 | 1020 | |||
GenBank mRNA | AK000505.1 | - | 8 | 999 | ||
AK304885.1 | - | 8 | 982 | |||
AL136677.1 | - | 8 | 977 | |||
BC012514.1 | - | 8 | 971 | 4509903 |