Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NOD1 SIRNA Assays ›
          Gene Symbol:
          NOD1
          Gene Name:
          nucleotide binding oligomerization domain containing 1
          Gene Aliases:
          CARD4, CLR7.1, NLRC1
          Chromosome Location:
          Chr.7: 30424527 - 30478840 on Build GRCh38
          UniGene ID:
          Hs.405153
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 135609

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NOD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006092.2 NP_006083.1 7, 8 2807
          XM_005249568.1 XP_005249625.1 8, 9 2862
          XM_005249572.1 XP_005249629.1 6, 7 2725
          XM_005249576.1 XP_005249633.1 3, 4 1624
          XM_006715633.2 XP_006715696.1 8, 9 2899
          XM_011515079.1 XP_011513381.1 9, 10 3045
          XM_011515080.2 XP_011513382.1 7, 8 2721
          XM_011515081.2 XP_011513383.1 7, 8 2871
          XM_011515083.1 XP_011513385.1 8, 9 2862
          XM_011515084.1 XP_011513386.1 8, 9 2862
          XM_011515085.1 XP_011513387.1 8, 9 2862
          XM_011515087.1 XP_011513389.1 8, 9 2862
          XM_017011674.1 XP_016867163.1 8, 9 2862
          XM_017011675.1 XP_016867164.1 7, 8 2768
          GenBank mRNA AF113925.1 - 7, 8 2708
          AF126484.1 - 7, 8 2735
          BC040339.1 - 7, 8 2722 4338930
          KU178514.1 - 4, 5 2284
          KU178515.1 - 3, 4 459
          KU178517.1 - 3, 4 459

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-motor microtubule binding protein scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPK activity
          positive regulation of dendritic cell antigen processing and presentation
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          defense response
          inflammatory response
          signal transduction
          JNK cascade
          detection of biotic stimulus
          detection of bacterium
          positive regulation of interleukin-1 beta production
          positive regulation of interleukin-6 production
          positive regulation of tumor necrosis factor production
          intracellular signal transduction
          interleukin-8 biosynthetic process
          defense response to bacterium
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          innate immune response
          positive regulation of JNK cascade
          defense response to Gram-positive bacterium
          positive regulation of nitric-oxide synthase activity
          positive regulation of NF-kappaB transcription factor activity
          protein oligomerization
          positive regulation of ERK1 and ERK2 cascade
          nucleotide-binding oligomerization domain containing signaling pathway
          cellular response to muramyl dipeptide
          positive regulation of NIK/NF-kappaB signaling
          positive regulation of xenophagy
          protein binding
          ATP binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          identical protein binding
          protein homodimerization activity
          peptidoglycan binding
          CARD domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qm47m:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0