Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CFDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006324.2 | NP_006315.1 | 7 | 1040 | ||
GenBank mRNA | AB009285.1 | - | 7 | 1012 | ||
BC000991.2 | - | 7 | 1036 | 3449836 | ||
D86549.1 | - | 6 | 839 |