Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Enterprise Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAP1 SIRNA Assays ›
Gene Symbol:
CAP1
Gene Name:
adenylate cyclase associated protein 1
Gene Aliases:
CAP, CAP1-PEN
Chromosome Location:
Chr.1: 40040070 - 40072649 on Build GRCh38
UniGene ID:
Hs.370581
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135704
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105530.1
NP_001099000.1
7
801
NM_006367.3
NP_006358.1
7
819
XM_005270368.1
XP_005270425.1
7
659
XM_011540509.1
XP_011538811.1
8
1212
XM_011540510.1
XP_011538812.1
8
1209
XM_011540511.1
XP_011538813.1
7
643
XM_011540512.1
XP_011538814.1
7
762
XM_011540513.2
XP_011538815.1
7
654
XM_011540514.1
XP_011538816.1
8
769
XM_011540515.1
XP_011538817.1
8
1209
XM_017000067.1
XP_016855556.1
7
727
XM_017000068.1
XP_016855557.1
8
771
XM_017000069.1
XP_016855558.1
8
982
XM_017000070.1
XP_016855559.1
7
1001
XM_017000071.1
XP_016855560.1
7
959
XM_017000072.1
XP_016855561.1
7
895
XM_017000073.1
XP_016855562.1
7
716
GenBank mRNA
AK096193.1
-
7
653
AK222513.1
-
7
660
AK295396.1
-
8
581
AK297829.1
-
7
528
AK298094.1
-
7
499
AK298110.1
-
7
603
AK300861.1
-
6
498
AK315731.1
-
7
706
BC013963.2
-
7
636
2963922
BC095440.1
-
7
641
30370256
BT007152.1
-
6
590
CR457409.1
-
6
590
L12168.1
-
6
629
M98474.1
-
7
660
Back To Top
More Information
Panther Classification:
Molecular Function -
actin or actin-binding cytoskeletal protein
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
ameboidal-type cell migration
receptor-mediated endocytosis
establishment or maintenance of cell polarity
signal transduction
activation of adenylate cyclase activity
actin polymerization or depolymerization
actin binding
adenylate cyclase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-qvstf:80/100.66.73.196:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0