Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SYNCRIP SIRNA Assays ›
          Gene Symbol:
          SYNCRIP
          Gene Name:
          synaptotagmin binding cytoplasmic RNA interacting protein
          Gene Aliases:
          GRY-RBP, GRYRBP, HNRNPQ, HNRPQ1, NSAP1, PP68, hnRNP-Q
          Chromosome Location:
          Chr.6: 85607784 - 85643862 on Build GRCh38
          UniGene ID:
          Hs.571177
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 135714

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SYNCRIP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001159673.1 NP_001153145.1 10 1273
          NM_001159674.1 NP_001153146.1 11 1296
          NM_001159675.1 NP_001153147.1 10 1202
          NM_001159676.1 NP_001153148.1 10 1307
          NM_001159677.1 NP_001153149.1 11 1836
          NM_001253771.1 NP_001240700.1 10 1215
          NM_006372.4 NP_006363.4 11 1836
          XM_005248635.4 XP_005248692.1 11 1457
          XM_005248636.2 XP_005248693.1 10 1687
          XM_005248637.2 XP_005248694.1 11 1420
          XM_017010176.1 XP_016865665.1 11 1457
          XM_017010177.1 XP_016865666.1 11 1457
          XM_017010178.1 XP_016865667.1 11 1417
          XM_017010179.1 XP_016865668.1 11 1392
          XM_017010180.1 XP_016865669.1 10 1472
          GenBank mRNA AB209098.1 - 10 1213
          AF037448.1 - 11 1820
          AF155568.1 - 11 1499
          AK222776.1 - 11 1445
          AK294212.1 - 4 506
          AK299796.1 - 10 1268
          AK313572.1 - 11 1456
          AY034481.1 - 10 1295
          AY034482.1 - 10 1190
          AY034483.1 - 10 1295
          BC032643.1 - 11 1287 5495201
          BC040844.1 - 10 1215 4513184

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          osteoblast differentiation
          RNA processing
          circadian rhythm
          RNA splicing
          viral process
          negative regulation of translation
          positive regulation of translation
          CRD-mediated mRNA stabilization
          cellular response to interferon-gamma
          negative regulation of mRNA modification
          nucleotide binding
          RNA binding
          mRNA 3'-UTR binding
          protein binding
          poly(A) binding
          poly(A) RNA binding
          mRNA 5'-UTR binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0