Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATG7 SIRNA Assays ›
Gene Symbol:
ATG7
Gene Name:
autophagy related 7
Gene Aliases:
APG7-LIKE, APG7L, GSA7
Chromosome Location:
Chr.3: 11272324 - 11564704 on Build GRCh38
UniGene ID:
Hs.38032
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
135754
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATG7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136031.2
NP_001129503.2
3
306
NM_001144912.1
NP_001138384.1
3
306
NM_006395.2
NP_006386.1
3
306
XM_006712931.3
XP_006712994.1
3
249
XM_006712932.3
XP_006712995.1
4
513
XM_006712933.3
XP_006712996.1
3
486
XM_011533277.2
XP_011531579.1
5
637
XM_011533278.2
XP_011531580.1
4
342
XM_011533279.2
XP_011531581.1
5
637
XM_011533280.2
XP_011531582.1
5
637
XM_011533281.2
XP_011531583.1
5
637
XM_011533282.2
XP_011531584.1
5
637
XM_011533283.2
XP_011531585.1
5
637
XM_011533284.2
XP_011531586.1
5
637
XM_011533285.2
XP_011531587.1
5
637
XM_011533286.2
XP_011531588.1
5
637
XM_017005542.1
XP_016861031.1
3
223
XM_017005543.1
XP_016861032.1
3
249
XM_017005544.1
XP_016861033.1
4
511
XM_017005545.1
XP_016861034.1
3
291
XM_017005546.1
XP_016861035.1
4
342
XM_017005547.1
XP_016861036.1
5
637
XM_017005548.1
XP_016861037.1
3
249
XM_017005549.1
XP_016861038.1
4
520
XM_017005550.1
XP_016861039.1
3
249
XM_017005551.1
XP_016861040.1
3
249
XM_017005552.1
XP_016861041.1
3
249
GenBank mRNA
AF094516.1
-
2
214
AK075221.1
-
3
233
AK298833.1
-
3
250
AK303694.1
-
3
233
AK314504.1
-
3
223
BC000091.2
-
3
221
3504204
DA177126.1
-
3
308
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
protein lipidation
C-terminal protein lipidation
cellular response to nitrogen starvation
cellular response to starvation
positive regulation of autophagy
protein transport
macroautophagy
positive regulation of macroautophagy
protein ubiquitination
positive regulation of protein modification process
piecemeal microautophagy of nucleus
suppression by virus of host autophagy
positive regulation of apoptotic process
late nucleophagy
positive regulation of protein catabolic process
defense response to virus
membrane fusion
cellular response to hyperoxia
regulation of mitophagy
ubiquitin activating enzyme activity
protein binding
transcription factor binding
Atg12 activating enzyme activity
Atg8 activating enzyme activity
protein homodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0