Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NUPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007342.2 | NP_031368.1 | 2 | 547 | ||
XM_005249592.1 | XP_005249649.1 | 2 | 410 | |||
XM_005249593.3 | XP_005249650.1 | 2 | 1090 | |||
XM_017011714.1 | XP_016867203.1 | 2 | 1090 | |||
XM_017011715.1 | XP_016867204.1 | 2 | 347 | |||
XM_017011716.1 | XP_016867205.1 | 2 | 1090 | |||
GenBank mRNA | AK298183.1 | - | 2 | 283 | ||
AK300113.1 | - | 2 | 336 | |||
AK312991.1 | - | 2 | 539 | |||
BC005327.1 | - | 2 | 350 | 3510844 | ||
BC107583.1 | - | 2 | 330 | 6453869 | ||
BT007409.1 | - | 2 | 288 | |||
DC398445.1 | - | 2 | 549 | |||
U97198.1 | - | 2 | 507 |