Search
Search

MAPRE1| Gene Symbol: MAPRE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012325.2 | NP_036457.1 | 5, 6 | 744 | ||
| XM_011528696.2 | XP_011526998.1 | 5, 6 | 746 | |||
| GenBank mRNA | AK297273.1 | - | 5, 6 | 593 | ||
| AK310817.1 | - | 2, 3 | 149 | |||
| AK312590.1 | - | 5, 6 | 661 | |||
| BC040410.1 | - | 5, 6 | 671 | |||
| BC106068.1 | - | 5, 6 | 674 | 6527006 | ||
| BC109281.3 | - | 5, 6 | 668 | 40031641 | ||
| U24166.1 | - | 5, 6 | 669 | |||