Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PASK SIRNA Assays ›
          Gene Symbol:
          PASK
          Gene Name:
          PAS domain containing serine/threonine kinase
          Gene Aliases:
          PASKIN, STK37
          Chromosome Location:
          Chr.2: 241106099 - 241150347 on Build GRCh38
          UniGene ID:
          Hs.397891
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 136787

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PASK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252119.1 NP_001239048.1 9 1520
          NM_001252120.1 NP_001239049.1 10 1673
          NM_001252122.1 NP_001239051.1 9 1415
          NM_001252124.1 NP_001239053.1 9 1520
          NM_015148.3 NP_055963.2 9 1520
          XM_005246991.1 XP_005247048.1 6 855
          XM_011510826.1 XP_011509128.1 9 1475
          XM_011510828.1 XP_011509130.1 10 1576
          XM_011510829.1 XP_011509131.1 9 1517
          XM_011510830.1 XP_011509132.1 9 1518
          XM_011510831.2 XP_011509133.1 7 1682
          XM_011510832.2 XP_011509134.1 7 1682
          XM_011510833.1 XP_011509135.1 6 855
          XM_011510834.1 XP_011509136.1 2 578
          XM_011510835.1 XP_011509137.1 2 168
          XM_017003634.1 XP_016859123.1 9 1764
          XM_017003635.1 XP_016859124.1 10 1576
          XM_017003636.1 XP_016859125.1 9 1532
          GenBank mRNA AF387103.1 - 9 1460
          AK056516.1 - 9 1465
          AK296945.1 - 6 1025
          AK302417.1 - 9 1371
          AL117400.1 - 1 1819
          BC020054.1 - 5 695
          BC050565.1 - 10 1479 4537404
          BC063585.1 - 9 1463 4419998
          CR749231.1 - 10 1674
          D50925.2 - 9 1451

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          regulation of respiratory gaseous exchange
          negative regulation of glycogen biosynthetic process
          positive regulation of translation
          protein autophosphorylation
          regulation of glucagon secretion
          regulation of energy homeostasis
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          phosphatidylinositol binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline