Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KANK1 SIRNA Assays ›
Gene Symbol:
KANK1
Gene Name:
KN motif and ankyrin repeat domains 1
Gene Aliases:
ANKRD15, CPSQ2, KANK
Chromosome Location:
Chr.9: 470294 - 746106 on Build GRCh38
UniGene ID:
Hs.306764
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
136800
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KANK1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001256876.1
NP_001243805.1
16
4698
NM_001256877.1
NP_001243806.1
13
4807
NM_015158.3
NP_055973.2
12
4192
NM_153186.4
NP_694856.1
11
4454
XM_005251411.2
XP_005251468.1
11
4775
XM_005251414.3
XP_005251471.1
10
5238
XM_005251415.2
XP_005251472.1
10
4967
XM_005251416.2
XP_005251473.1
10
4743
XM_005251417.3
XP_005251474.1
10
5234
XM_005251418.2
XP_005251475.1
10
5245
XM_005251419.2
XP_005251476.1
10
4936
XM_006716743.2
XP_006716806.1
11
4751
XM_011517819.1
XP_011516121.1
12
4866
XM_011517820.1
XP_011516122.1
11
4883
XM_011517821.1
XP_011516123.1
11
4810
XM_017014511.1
XP_016870000.1
12
4988
XM_017014512.1
XP_016870001.1
11
6299
XM_017014513.1
XP_016870002.1
13
5501
XM_017014514.1
XP_016870003.1
12
4604
XM_017014515.1
XP_016870004.1
13
4340
XM_017014516.1
XP_016870005.1
11
5133
XM_017014517.1
XP_016870006.1
12
4550
XM_017014518.1
XP_016870007.1
12
4138
XM_017014519.1
XP_016870008.1
10
4947
XM_017014520.1
XP_016870009.1
10
4535
XM_017014521.1
XP_016870010.1
11
4364
XM_017014522.1
XP_016870011.1
12
4165
XM_017014523.1
XP_016870012.1
11
3952
XM_017014524.1
XP_016870013.1
10
4893
XM_017014525.1
XP_016870014.1
11
4310
XM_017014526.1
XP_016870015.1
11
5220
XM_017014527.1
XP_016870016.1
11
4384
XM_017014528.1
XP_016870017.1
12
4238
XM_017014529.1
XP_016870018.1
11
4651
XM_017014530.1
XP_016870019.1
11
4160
XM_017014531.1
XP_016870020.1
10
4913
XM_017014532.1
XP_016870021.1
10
5187
XM_017014533.1
XP_016870022.1
10
4684
XM_017014534.1
XP_016870023.1
11
4330
XM_017014535.1
XP_016870024.1
11
4596
XM_017014536.1
XP_016870025.1
11
4299
XM_017014537.1
XP_016870026.1
9
4516
XM_017014538.1
XP_016870027.1
9
5001
XM_017014539.1
XP_016870028.1
10
3933
XM_017014540.1
XP_016870029.1
10
4409
XM_017014541.1
XP_016870030.1
10
4413
XM_017014542.1
XP_016870031.1
10
3866
GenBank mRNA
AK025428.1
-
10
2337
AK025636.1
-
5
1389
AK290028.1
-
12
4158
AK292989.1
-
12
4171
AL833015.1
-
8
1104
AL833161.1
-
12
4172
BC020040.1
-
10
2346
BC037495.2
-
16
4701
5265517
BC038116.2
-
11
4364
5261060
BX648931.1
-
11
6318
D79994.2
-
12
4757
Back To Top
More Information
Panther Classification:
Molecular Function -
non-motor actin binding protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
negative regulation of neuron projection development
positive regulation of Wnt signaling pathway
negative regulation of cell migration
negative regulation of actin filament polymerization
negative regulation of Rho protein signal transduction
positive regulation of catenin import into nucleus
negative regulation of insulin receptor signaling pathway
positive regulation of wound healing
negative regulation of substrate adhesion-dependent cell spreading
negative regulation of ruffle assembly
regulation of establishment of cell polarity
negative regulation of lamellipodium morphogenesis
protein binding
beta-catenin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System