Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Pipettes and Pipette Tips
Lab Centrifuges
Ultra-Low Temperature Freezers
Spectroscopy
Beakers
PCR Equipment and Supplies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
See all product categories
Applications
Cell Analysis
Lab Equipment
Real-Time PCR
PCR
Chromatography
Cell Culture and Transfection
DNA and RNA Extraction and Analysis
Protein Biology
Flow Cytometry
Chemicals
See all applications and techniques
Services
Custom Services
Lab Informatics
Enterprise Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Instrument Services
Training Services
See all services
Help and Support
Customer Center
Contact Us
Certificates of Analysis and Conformance
Safety Data Sheets (SDS)
Manuals
How to Cite Our Products in a Paper
Instrument Support
Knowledge Base and Product FAQs
Learning Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SYNE1 SIRNA Assays ›
Gene Symbol:
SYNE1
Gene Name:
spectrin repeat containing nuclear envelope protein 1
Gene Aliases:
8B, ARCA1, C6orf98, CPG2, EDMD4, MYNE1, Nesp1, SCAR8, dJ45H2.2
Chromosome Location:
Chr.6: 152121684 - 152637399 on Build GRCh38
UniGene ID:
Hs.12967
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
137093
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SYNE1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_033071.3
NP_149062.1
129
23806
NM_182961.3
NP_892006.3
130
24184
XM_006715407.1
XP_006715470.1
130
24097
XM_006715408.2
XP_006715471.1
130
24089
XM_006715409.1
XP_006715472.1
129
24076
XM_006715410.2
XP_006715473.1
130
24101
XM_006715411.1
XP_006715474.1
129
24046
XM_006715412.2
XP_006715475.1
130
24086
XM_006715413.2
XP_006715476.1
130
24101
XM_006715414.1
XP_006715477.1
128
24025
XM_006715415.2
XP_006715478.1
130
24101
XM_006715416.2
XP_006715479.1
130
24086
XM_006715417.2
XP_006715480.1
129
23960
XM_006715420.2
XP_006715483.1
129
23948
XM_006715421.2
XP_006715484.1
129
23945
XM_006715422.1
XP_006715485.1
129
23938
XM_006715423.2
XP_006715486.1
130
24101
XM_006715424.2
XP_006715487.1
130
24101
XM_006715425.2
XP_006715488.1
130
24101
XM_011535641.2
XP_011533943.1
130
24098
XM_011535642.2
XP_011533944.1
130
24086
XM_011535643.1
XP_011533945.1
129
23932
XM_011535644.1
XP_011533946.1
116
22148
XM_011535645.2
XP_011533947.1
112
21541
XM_017010608.1
XP_016866097.1
131
24216
XM_017010609.1
XP_016866098.1
130
24023
XM_017010610.1
XP_016866099.1
129
24002
XM_017010611.1
XP_016866100.1
129
23805
XM_017010612.1
XP_016866101.1
128
23729
XM_017010613.1
XP_016866102.1
130
24098
XM_017010614.1
XP_016866103.1
129
23945
XM_017010615.1
XP_016866104.1
129
23945
XM_017010616.1
XP_016866105.1
130
24101
XM_017010617.1
XP_016866106.1
130
24098
XM_017010618.1
XP_016866107.1
130
24086
XM_017010619.1
XP_016866108.1
116
22180
GenBank mRNA
AB018339.2
-
18
2821
AF444779.1
-
5
694
AF495910.1
-
130
24184
AF535142.1
-
129
23806
AK122691.1
-
10
1464
AK127076.1
-
14
2161
AK304825.1
-
3
386
AK316101.1
-
3
401
AY061755.1
-
45
7274
AY061756.1
-
2
264
AY184206.1
-
43
7169
BC090927.1
-
3
446
BX537837.1
-
12
1762
BX647837.1
-
15
2356
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
nucleus organization
Golgi organization
establishment of nucleus localization
muscle cell differentiation
cytoskeletal anchoring at nuclear membrane
nuclear matrix anchoring at nuclear membrane
actin binding
protein binding
lamin binding
protein homodimerization activity
poly(A) RNA binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0