Search
Search

CD101| Gene Symbol: CD101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256106.2 | NP_001243035.1 | 3 | 826 | ||
| NM_001256109.2 | NP_001243038.1 | 3 | 826 | |||
| NM_001256111.2 | NP_001243040.1 | 3 | 640 | |||
| NM_004258.5 | NP_004249.2 | 3 | 826 | |||
| XM_017002846.1 | XP_016858335.1 | 4 | 779 | |||
| XM_017002847.1 | XP_016858336.1 | 4 | 779 | |||
| GenBank mRNA | AK298237.1 | - | 3 | 615 | ||
| AK307542.1 | - | 3 | 826 | |||
| BC130327.1 | - | 3 | 787 | 40146358 | ||
| BC136484.1 | - | 3 | 787 | 9020473 | ||
| BC171768.1 | - | 3 | 787 | 9054422 | ||
| Z33642.1 | - | 3 | 779 | |||