Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: BUB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007793.2 | NP_001007794.1 | 5, 6 | 823 | ||
NM_004725.3 | NP_004716.1 | 5, 6 | 823 | |||
GenBank mRNA | AF047472.1 | - | 5, 6 | 651 | ||
AF047473.1 | - | 5, 6 | 651 | |||
AF053304.1 | - | 5, 6 | 619 | |||
AF081496.1 | - | 5, 6 | 668 | |||
AK226060.1 | - | 2406 | ||||
AK293255.1 | - | 4, 5 | 479 | |||
AK307958.1 | - | 5, 6 | 645 | |||
AK312546.1 | - | 5, 6 | 691 | |||
BC005138.2 | - | 5, 6 | 652 | 3504520 | ||
BC022438.1 | - | 5, 6 | 644 | 4281970 | ||
BX647392.1 | - | 5, 6 | 700 |