Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PAPSS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005443.4 | NP_005434.4 | 11 | 1802 | ||
XM_011532400.1 | XP_011530702.1 | 11 | 1782 | |||
XM_011532401.1 | XP_011530703.1 | 11 | 1868 | |||
GenBank mRNA | AF016496.1 | - | 11 | 1718 | ||
AF033026.1 | - | 11 | 1744 | |||
AF105227.1 | - | 11 | 1772 | |||
AK292774.1 | - | 11 | 1752 | |||
BC011392.1 | - | 11 | 1675 | 3869484 | ||
BC040940.1 | - | 11 | 1730 | |||
BC050627.1 | - | 11 | 1738 | 5556165 | ||
BU517185.1 | - | 1 | 66 | |||
CR457028.1 | - | 11 | 1718 | |||
Y10387.1 | - | 11 | 1754 |