Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005772.4 | NP_005763.3 | 3 | 621 | ||
XM_006716715.3 | XP_006716778.1 | 3 | 568 | |||
GenBank mRNA | AF067172.1 | - | 3 | 340 | ||
AF161456.1 | - | 2 | 230 | |||
AJ276894.1 | - | 3 | 363 | |||
AK022904.1 | - | 3 | 518 | |||
AK225872.1 | - | 3 | 532 | |||
BC001025.2 | - | 3 | 514 | 3343468 |