Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ST18 SIRNA Assays ›
Gene Symbol:
ST18
Gene Name:
ST18, C2H2C-type zinc finger
Gene Aliases:
NZF3, ZC2H2C3, ZC2HC10, ZNF387
Chromosome Location:
Chr.8: 52110832 - 52409879 on Build GRCh38
UniGene ID:
Hs.655499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
138328
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ST18
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_014682.2
NP_055497.1
10
1318
XM_006716487.1
XP_006716550.1
10
1085
XM_011517629.1
XP_011515931.1
11
1241
XM_011517631.1
XP_011515933.1
12
1282
XM_011517632.1
XP_011515934.1
10
1096
XM_011517633.1
XP_011515935.1
10
1089
XM_011517634.1
XP_011515936.1
10
1285
XM_011517635.1
XP_011515937.1
10
1289
XM_011517636.2
XP_011515938.1
9
1366
XM_011517637.1
XP_011515939.1
8
823
XM_011517638.2
XP_011515940.1
7
864
XM_011517641.1
XP_011515943.1
12
1329
XM_011517642.1
XP_011515944.1
11
1241
XM_017014047.1
XP_016869536.1
10
1445
XM_017014048.1
XP_016869537.1
10
1449
XM_017014049.1
XP_016869538.1
11
1495
XM_017014050.1
XP_016869539.1
10
1282
XM_017014051.1
XP_016869540.1
9
1289
XM_017014052.1
XP_016869541.1
11
1499
XM_017014053.1
XP_016869542.1
11
1486
XM_017014054.1
XP_016869543.1
9
1535
XM_017014055.1
XP_016869544.1
9
1126
XM_017014056.1
XP_016869545.1
12
1536
XM_017014057.1
XP_016869546.1
10
1339
XM_017014058.1
XP_016869547.1
10
1100
XM_017014059.1
XP_016869548.1
10
1286
XM_017014060.1
XP_016869549.1
11
1323
XM_017014061.1
XP_016869550.1
9
1458
XM_017014062.1
XP_016869551.1
8
1210
XM_017014063.1
XP_016869552.1
9
1756
XM_017014064.1
XP_016869553.1
9
1370
XM_017014065.1
XP_016869554.1
11
1490
XM_017014066.1
XP_016869555.1
9
1129
XM_017014067.1
XP_016869556.1
9
1539
XM_017014068.1
XP_016869557.1
10
1330
XM_017014069.1
XP_016869558.1
7
1027
XM_017014070.1
XP_016869559.1
6
948
XM_017014071.1
XP_016869560.1
7
1196
XM_017014072.1
XP_016869561.1
10
1447
XM_017014073.1
XP_016869562.1
10
1085
XM_017014074.1
XP_016869563.1
9
1368
XM_017014075.1
XP_016869564.1
11
1532
XM_017014076.1
XP_016869565.1
11
1536
XM_017014077.1
XP_016869566.1
11
1173
XM_017014078.1
XP_016869567.1
10
1453
XM_017014079.1
XP_016869568.1
10
1844
XM_017014080.1
XP_016869569.1
8
1085
XM_017014081.1
XP_016869570.1
10
1452
XM_017014082.1
XP_016869571.1
10
1287
XM_017014083.1
XP_016869572.1
9
1127
XM_017014084.1
XP_016869573.1
7
865
GenBank mRNA
AB011107.1
-
10
1182
BC031075.1
-
10
1215
BC117147.1
-
5
575
40125698
BC117149.1
-
5
575
40125700
BC143872.1
-
5
570
BC143873.1
-
6
663
Back To Top
More Information
Panther Classification:
Molecular Function -
zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0