Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TIAM1 SIRNA Assays ›
          Gene Symbol:
          TIAM1
          Gene Name:
          T-cell lymphoma invasion and metastasis 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.21: 31118418 - 31559977 on Build GRCh38
          UniGene ID:
          Hs.517228
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 138877

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TIAM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003253.2 NP_003244.2 5 778
          XM_005261037.1 XP_005261094.1 4 911
          XM_005261038.2 XP_005261095.1 3 645
          XM_005261040.2 XP_005261097.1 4 911
          XM_011529711.1 XP_011528013.1 5 862
          XM_011529712.1 XP_011528014.1 5 786
          XM_011529713.1 XP_011528015.1 5 1196
          XM_017028447.1 XP_016883936.1 5 965
          XM_017028448.1 XP_016883937.1 5 935
          XM_017028449.1 XP_016883938.1 4 842
          XM_017028450.1 XP_016883939.1 4 862
          XM_017028451.1 XP_016883940.1 4 1342
          XM_017028452.1 XP_016883941.1 6 1142
          XM_017028453.1 XP_016883942.1 4 817
          GenBank mRNA BC117192.1 - 2 353 40125743
          BC117196.1 - 2 353 40125747
          BC143970.1 - 2 353
          BC143980.1 - 2 353 9052498
          U16296.1 - 5 779

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cardiac muscle hypertrophy
          apoptotic process
          cell-matrix adhesion
          cell migration
          Rac protein signal transduction
          positive regulation of protein binding
          regulation of Rho protein signal transduction
          response to cocaine
          positive regulation of apoptotic process
          positive regulation of JUN kinase activity
          ephrin receptor signaling pathway
          positive regulation of axonogenesis
          regulation of small GTPase mediated signal transduction
          Wnt signaling pathway, planar cell polarity pathway
          positive regulation of dendritic spine morphogenesis
          regulation of insulin secretion involved in cellular response to glucose stimulus
          regulation of ERK1 and ERK2 cascade
          protein localization to membrane
          activation of GTPase activity
          positive regulation of Schwann cell chemotaxis
          regulation of dopaminergic neuron differentiation
          neuron projection extension
          regulation of non-canonical Wnt signaling pathway
          receptor signaling protein activity
          guanyl-nucleotide exchange factor activity
          Rho guanyl-nucleotide exchange factor activity
          protein binding
          microtubule binding
          lipid binding
          kinase binding
          Rac guanyl-nucleotide exchange factor activity
          receptor tyrosine kinase binding
          Rac GTPase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0