Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFX SIRNA Assays ›
Gene Symbol:
ZFX
Gene Name:
zinc finger protein, X-linked
Gene Aliases:
ZNF926
Chromosome Location:
Chr.X: 24148994 - 24216255 on Build GRCh38
UniGene ID:
Hs.336681
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139034
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001178084.1
NP_001171555.1
3
511
NM_001178085.1
NP_001171556.1
6
855
NM_001178095.1
NP_001171566.1
3
590
NM_003410.3
NP_003401.2
5
719
XM_005274591.4
XP_005274648.1
4
885
XM_005274592.3
XP_005274649.1
3
700
XM_005274594.4
XP_005274651.1
2
1890
XM_006724513.3
XP_006724576.1
6
1114
XM_006724515.3
XP_006724578.1
6
1116
XM_011545578.1
XP_011543880.1
6
996
XM_011545579.1
XP_011543881.1
5
784
XM_011545581.2
XP_011543883.1
5
926
XM_011545583.2
XP_011543885.1
2
440
XM_017029788.1
XP_016885277.1
6
1037
XM_017029789.1
XP_016885278.1
5
1599
XM_017029790.1
XP_016885279.1
5
1004
XM_017029791.1
XP_016885280.1
4
894
XM_017029792.1
XP_016885281.1
4
773
XM_017029793.1
XP_016885282.1
3
571
XM_017029794.1
XP_016885283.1
5
825
XM_017029795.1
XP_016885284.1
2
454
XM_017029796.1
XP_016885285.1
4
606
XM_017029797.1
XP_016885286.1
6
1065
XM_017029798.1
XP_016885287.1
2
685
XM_017029799.1
XP_016885288.1
3
700
XM_017029800.1
XP_016885289.1
2
687
GenBank mRNA
AB209892.1
-
3
623
AF505615.1
-
2
412
AF505616.1
-
2
412
AF505618.1
-
2
412
AF505619.1
-
2
412
BC136308.1
-
5
707
9020295
BC136309.1
-
5
707
9020296
DA995443.1
-
3
513
M30608.1
-
4
670
X59739.1
-
3
490
X59740.1
-
6
839
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
RNA polymerase II regulatory region sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
transcription coactivator activity
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0