Search
Search

BCAS1| Gene Symbol: BCAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003657.3 | NP_003648.2 | 9 | 1567 | ||
| XM_005260587.2 | XP_005260644.1 | 10 | 1482 | |||
| XM_005260589.2 | XP_005260646.1 | 9 | 1440 | |||
| XM_005260590.2 | XP_005260647.1 | 10 | 1482 | |||
| XM_005260591.2 | XP_005260648.1 | 9 | 1440 | |||
| XM_005260593.2 | XP_005260650.1 | 8 | 1305 | |||
| XM_011529090.2 | XP_011527392.1 | 10 | 1410 | |||
| XM_017028110.1 | XP_016883599.1 | 9 | 1347 | |||
| XM_017028111.1 | XP_016883600.1 | 8 | 1305 | |||
| GenBank mRNA | AF041260.1 | - | 9 | 1346 | ||
| AK225656.1 | - | 8 | 1294 | |||
| AK294135.1 | - | 5 | 466 | |||
| AK295318.1 | - | 7 | 881 | |||
| AK315220.1 | - | 9 | 1567 | |||
| BC126346.1 | - | 9 | 1278 | 8992062 | ||
| BC126348.1 | - | 9 | 1278 | 8992064 | ||
| CR749643.1 | - | 5 | 502 | |||