Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PRKRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139517.1 | NP_001132989.1 | 2 | 530 | ||
NM_001139518.1 | NP_001132990.1 | 3 | 251 | |||
NM_001316362.1 | NP_001303291.1 | 3 | 459 | |||
NM_003690.4 | NP_003681.1 | 3 | 450 | |||
XM_011512063.1 | XP_011510365.1 | 3 | 332 | |||
XM_011512066.1 | XP_011510368.1 | 3 | 496 | |||
XM_017005159.1 | XP_016860648.1 | 3 | 332 | |||
GenBank mRNA | AF072860.1 | - | 3 | 345 | ||
AF083033.1 | - | 3 | 341 | |||
AK223107.1 | - | 3 | 349 | |||
AK290601.1 | - | 3 | 376 | |||
AK296019.1 | - | 3 | 370 | |||
AL136615.1 | - | 3 | 374 | |||
AL833867.1 | - | 2 | 530 | |||
AY251164.1 | - | 3 | 268 | |||
BC009470.1 | - | 3 | 356 | 3503747 | ||
BG827125.1 | - | 3 | 451 | |||
BT007243.1 | - | 3 | 249 | |||
CR533525.1 | - | 3 | 249 |