Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RUVBL1 SIRNA Assays ›
          Gene Symbol:
          RUVBL1
          Gene Name:
          RuvB like AAA ATPase 1
          Gene Aliases:
          ECP-54, ECP54, INO80H, NMP 238, NMP238, PONTIN, Pontin52, RVB1, TIH1, TIP49, TIP49A
          Chromosome Location:
          Chr.3: 128064612 - 128153914 on Build GRCh38
          UniGene ID:
          Hs.272822
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 139337

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RUVBL1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001319084.1 NP_001306013.1 9 1259
          NM_001319086.1 NP_001306015.1 9 1667
          NM_003707.2 NP_003698.1 9 1200
          XM_005247841.3 XP_005247898.1 9 1121
          XM_011513248.1 XP_011511550.1 10 1354
          XM_011513249.2 XP_011511551.1 9 1254
          XM_017007356.1 XP_016862845.1 9 1254
          XM_017007357.1 XP_016862846.1 9 1124
          GenBank mRNA AB012122.1 - 9 1149
          AB451224.1 - 9 1096
          AB451347.1 - 9 1096
          AF070735.1 - 9 1172
          AF099084.1 - 9 1160
          AJ010058.1 - 9 1172
          AK092163.1 - 9 1667
          AK222563.1 - 9 1181
          AK222575.1 - 9 1172
          AK312290.1 - 9 1196
          BC002993.1 - 9 1167 2823568
          BC012886.1 - 9 1129 3876400
          BT007057.1 - 9 1096
          DQ469310.1 - 9 1096
          Y18418.1 - 9 1192

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          box C/D snoRNP assembly
          DNA repair
          DNA recombination
          chromatin remodeling
          transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          mitotic nuclear division
          spermatogenesis
          DNA duplex unwinding
          CENP-A containing nucleosome assembly
          regulation of growth
          histone H4 acetylation
          histone H2A acetylation
          cell division
          cell-cell adhesion
          regulation of mitophagy
          positive regulation of protein targeting to mitochondrion
          beta-catenin-TCF complex assembly
          positive regulation of telomerase RNA localization to Cajal body
          DNA helicase activity
          protein binding
          ATP binding
          ATPase activity
          ATP-dependent 5'-3' DNA helicase activity
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0