Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SSPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135823.1 | NP_001129295.1 | 2 | 261 | ||
NM_005086.4 | NP_005077.2 | 2 | 444 | |||
XM_011520853.2 | XP_011519155.1 | 2 | 194 | |||
XM_011520854.2 | XP_011519156.1 | 2 | 368 | |||
XM_011520855.2 | XP_011519157.1 | 2 | 110 | |||
GenBank mRNA | AF016028.2 | - | 2 | 344 | ||
AK092925.1 | - | 2 | 257 | |||
AK304667.1 | - | 3 | 338 | |||
AK314353.1 | - | 2 | 433 | |||
AL136756.1 | - | 2 | 428 | |||
BC062299.1 | - | 2 | 424 | 6376868 | ||
BP341300.1 | - | 2 | 444 | |||
DA073350.1 | - | 2 | 261 | |||
EU433933.1 | - | 2 | 344 | |||
X89105.1 | - | 2 | 266 |