Search
Search

COLQ| Gene Symbol: COLQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005677.3 | NP_005668.2 | 17 | 2682 | ||
| NM_080538.2 | NP_536799.1 | 17 | 2811 | |||
| NM_080539.3 | NP_536800.2 | 16 | 2580 | |||
| GenBank mRNA | AF057036.1 | - | 17 | 2682 | ||