Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TACC1 SIRNA Assays ›
Gene Symbol:
TACC1
Gene Name:
transforming acidic coiled-coil containing protein 1
Gene Aliases:
Ga55
Chromosome Location:
Chr.8: 38728186 - 38853028 on Build GRCh38
UniGene ID:
Hs.279245
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139591
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TACC1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146216.2
NP_001139688.1
3
804
NM_006283.2
NP_006274.2
3
858
XM_005273625.4
XP_005273682.1
4
972
XM_005273628.1
XP_005273685.1
3
479
XM_005273629.1
XP_005273686.1
4
492
XM_005273630.1
XP_005273687.1
3
500
XM_011544631.1
XP_011542933.1
3
858
XM_011544632.2
XP_011542934.1
5
1033
XM_011544633.1
XP_011542935.1
3
858
XM_011544634.1
XP_011542936.1
3
858
XM_011544635.1
XP_011542937.1
3
562
XM_017013766.1
XP_016869255.1
3
479
XM_017013767.1
XP_016869256.1
4
492
XM_017013768.1
XP_016869257.1
3
479
XM_017013769.1
XP_016869258.1
4
492
XM_017013770.1
XP_016869259.1
2
392
XM_017013771.1
XP_016869260.1
2
664
XM_017013772.1
XP_016869261.1
2
392
XM_017013773.1
XP_016869262.1
2
659
XM_017013774.1
XP_016869263.1
2
344
XM_017013775.1
XP_016869264.1
3
500
XM_017013776.1
XP_016869265.1
4
492
XM_017013777.1
XP_016869266.1
2
656
XM_017013778.1
XP_016869267.1
2
355
GenBank mRNA
AF049910.1
-
3
799
AK294931.1
-
3
459
AK295071.1
-
3
406
AK295841.1
-
2
395
AK303596.1
-
2
290
AK304507.1
-
5
691
AK314620.1
-
3
858
AY072876.2
-
3
410
BC041391.1
-
5
1055
5276767
BX647696.1
-
3
806
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
cell cycle
cell proliferation
cerebral cortex development
cell division
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0