Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: DYNLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291602.1 | NP_001278531.1 | 4 | 515 | ||
NM_001291603.1 | NP_001278532.1 | 5 | 537 | |||
NM_006519.3 | NP_006510.1 | 5 | 557 | |||
GenBank mRNA | AK026669.1 | - | 2 | 2681 | ||
BC029412.1 | - | 5 | 510 | 4074224 | ||
BC105588.1 | - | 5 | 506 | 6723966 | ||
D50663.1 | - | 5 | 491 | |||
HY017024.1 | - | 4 | 468 | |||
U56255.1 | - | 5 | 479 |