Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TRO SIRNA Assays ›
Gene Symbol:
TRO
Gene Name:
trophinin
Gene Aliases:
MAGE-d3, MAGED3
Chromosome Location:
Chr.X: 54920563 - 54931436 on Build GRCh38
UniGene ID:
Hs.633653
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139808
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TRO
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001039705.2
NP_001034794.1
13
4793
NM_001271183.1
NP_001258112.1
11
3513
NM_001271184.1
NP_001258113.1
12
3413
NM_016157.3
NP_057241.2
13
2472
NM_177556.2
NP_808224.1
13
2457
NM_177557.2
NP_808225.1
12
1266
NR_073148.1
14
2748
NR_073149.1
13
1557
XM_006724600.2
XP_006724663.1
13
4559
XM_011530808.1
XP_011529110.1
13
4604
XM_011530809.1
XP_011529111.1
13
4555
XM_011530811.2
XP_011529113.1
13
4941
XM_011530812.1
XP_011529114.1
13
4542
XM_011530813.1
XP_011529115.1
13
4592
XM_011530814.1
XP_011529116.1
13
4486
XM_017029767.1
XP_016885256.1
13
4956
XM_017029768.1
XP_016885257.1
13
4573
XM_017029769.1
XP_016885258.1
13
2234
XM_017029770.1
XP_016885259.1
13
2283
XM_017029771.1
XP_016885260.1
13
2611
XM_017029772.1
XP_016885261.1
13
2252
XM_017029773.1
XP_016885262.1
13
2163
GenBank mRNA
AB029037.2
-
13
4514
AF349719.1
-
14
2495
AF349720.1
-
13
2204
AF349721.1
-
13
1303
AF349722.1
-
12
1013
AK001300.1
-
11
1902
AK027489.1
-
13
2211
AK027861.1
-
9
3577
AK225841.1
-
12
3386
AK296329.1
-
11
3252
AK299568.1
-
4170
BC026914.1
-
14
2478
BM693634.1
-
3
490
U04811.2
-
2
2389
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
homophilic cell adhesion via plasma membrane adhesion molecules
embryo implantation
negative regulation of cell growth
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-7l2qm:80/100.66.75.127:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline