Search
Search

TARS| Gene Symbol: TARS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001258437.1 | NP_001245366.1 | 19 | 2108 | ||
| NM_001258438.1 | NP_001245367.1 | 19 | 2381 | |||
| NM_152295.4 | NP_689508.3 | 18 | 2282 | |||
| NR_047676.1 | 19 | 2403 | ||||
| NR_047677.1 | 17 | 2121 | ||||
| NR_047678.1 | 17 | 2091 | ||||
| GenBank mRNA | AK095852.1 | - | 17 | 1903 | ||
| AK222804.1 | - | 18 | 2039 | |||
| AK292346.1 | - | 19 | 2108 | |||
| AK293620.1 | - | 19 | 2192 | |||
| AK296783.1 | - | 17 | 1920 | |||
| AK315713.1 | - | 18 | 2093 | |||
| BC000517.2 | - | 18 | 2051 | 2822851 | ||
| BC003699.1 | - | 2 | 83 | |||
| BC010578.1 | - | 18 | 2069 | 3458845 | ||
| BC088355.1 | - | 18 | 2060 | |||
| BG183397.1 | - | 1 | 67 | |||
| M63180.1 | - | 18 | 2076 | |||