Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LETMD1 SIRNA Assays ›
          Gene Symbol:
          LETMD1
          Gene Name:
          LETM1 domain containing 1
          Gene Aliases:
          1110019O13Rik, HCCR-1, HCCR-2, HCCR1, HCRR-2
          Chromosome Location:
          Chr.12: 51048230 - 51068984 on Build GRCh38
          UniGene ID:
          Hs.655272
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 140259

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LETMD1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243689.1 NP_001230618.1 9 1526
          NM_001300765.1 NP_001287694.1 6 1115
          NM_015416.4 NP_056231.3 9 1487
          NR_045017.1 9 1362
          NR_045018.1 8 1371
          NR_045019.1 7 1288
          NR_045020.1 5 999
          XM_006719335.1 XP_006719398.1 8 1309
          XM_006719336.1 XP_006719399.1 8 1197
          XM_006719339.1 XP_006719402.1 9 1425
          XM_006719340.1 XP_006719403.1 8 1246
          XM_011538161.2 XP_011536463.1 9 1494
          XM_011538162.2 XP_011536464.1 8 1398
          XM_011538164.2 XP_011536466.1 8 1344
          XM_011538165.2 XP_011536467.1 6 1122
          XM_011538167.1 XP_011536469.1 4 916
          XM_017019163.1 XP_016874652.1 7 1156
          XM_017019167.1 XP_016874656.1 5 990
          GenBank mRNA AF195651.2 - 9 1459
          AF315598.1 - 9 1323
          AF329277.1 - 8 1370
          AK000639.1 - 9 1474
          AK057866.1 - 9 1432
          AK123080.1 - 1 2305
          AK126372.1 - 1 1597
          AK127540.1 - 9 1487
          AK225836.1 - 9 1445
          AK301955.1 - 9 1362
          AL050286.1 - 4 979
          BC000395.2 - 4 736
          BC019274.1 - 4 736
          BC064943.1 - 9 1433 6009854

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          secondary carrier transporter

          Gene Ontology Categories:

          Process(es)

          protein binding
          ribosome binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0