Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CDK12 SIRNA Assays ›
          Gene Symbol:
          CDK12
          Gene Name:
          cyclin dependent kinase 12
          Gene Aliases:
          CRK7, CRKR, CRKRS
          Chromosome Location:
          Chr.17: 39461486 - 39567560 on Build GRCh38
          UniGene ID:
          Hs.345028
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 140363

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CDK12
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_015083.2 NP_055898.1 1 692
          NM_016507.3 NP_057591.2 1 692
          XM_005257456.3 XP_005257513.1 1 717
          XM_005257458.4 XP_005257515.1 1 717
          XM_011524892.2 XP_011523194.1 1 717
          XM_011524893.2 XP_011523195.1 1 717
          XM_011524894.2 XP_011523196.1 1 717
          XM_011524895.2 XP_011523197.1 1 717
          XM_011524896.2 XP_011523198.1 1 717
          XM_011524897.2 XP_011523199.1 1 717
          XM_011524898.2 XP_011523200.1 1 717
          XM_011524899.2 XP_011523201.1 1 717
          XM_011524900.2 XP_011523202.1 1 717
          XM_011524901.2 XP_011523203.1 1 717
          XM_011524902.2 XP_011523204.1 1 717
          XM_011524903.2 XP_011523205.1 1 717
          XM_011524905.2 XP_011523207.1 1 717
          XM_011524906.2 XP_011523208.1 1 717
          XM_011524907.2 XP_011523209.1 1 717
          XM_017024744.1 XP_016880233.1 1 717
          XM_017024745.1 XP_016880234.1 1 717
          XM_017024746.1 XP_016880235.1 1 717
          XM_017024747.1 XP_016880236.1 1 717
          XM_017024748.1 XP_016880237.1 1 717
          XM_017024749.1 XP_016880238.1 1 717
          XM_017024750.1 XP_016880239.1 1 717
          XM_017024751.1 XP_016880240.1 1 717
          XM_017024752.1 XP_016880241.1 1 717
          XM_017024753.1 XP_016880242.1 1 717
          GenBank mRNA AB020711.2 - 1 442
          AF227198.1 - 1 164
          AK302645.1 - 1 692
          BC140854.1 - 1 151 9021722
          BC150265.1 - 1 448 9007217

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_138916, NM_001033867, XM_006247250, XM_006247251, XM_006247253, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001109626, NM_026952, NM_001109628, BC116645, AK007584, and others.

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA processing
          RNA splicing
          regulation of MAP kinase activity
          regulation of RNA splicing
          protein autophosphorylation
          regulation of cell cycle
          phosphorylation of RNA polymerase II C-terminal domain
          protein kinase activity
          cyclin-dependent protein serine/threonine kinase activity
          protein binding
          ATP binding
          RNA polymerase II carboxy-terminal domain kinase activity
          cyclin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0