Search
Search

OSMR| Gene Symbol: OSMR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168355.2 | NP_001161827.1 | 4, 5 | 698 | ||
| NM_001323504.1 | NP_001310433.1 | 4, 5 | 818 | |||
| NM_001323505.1 | NP_001310434.1 | 4, 5 | 698 | |||
| NM_001323506.1 | NP_001310435.1 | 4, 5 | 698 | |||
| NM_001323507.1 | NP_001310436.1 | 4, 5 | 698 | |||
| NM_003999.2 | NP_003990.1 | 4, 5 | 818 | |||
| XM_005248384.1 | XP_005248441.1 | 4, 5 | 805 | |||
| XM_005248386.2 | XP_005248443.1 | 4, 5 | 805 | |||
| XM_005248387.2 | XP_005248444.1 | 4, 5 | 805 | |||
| XM_011514161.2 | XP_011512463.1 | 4, 5 | 805 | |||
| XM_017010019.1 | XP_016865508.1 | 4, 5 | 805 | |||
| GenBank mRNA | BC010943.1 | - | 4, 5 | 548 | ||
| BC063468.1 | - | 4, 5 | 470 | |||
| BC125209.1 | - | 4, 5 | 588 | 40123501 | ||
| BC125210.1 | - | 4, 5 | 588 | 40123502 | ||
| U60805.1 | - | 4, 5 | 783 | |||